ContactUsLinkComponent
Rat FGF6 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat FGF6 in serum, plasma or other biological fluids. 1 * 96 Tests
  ANTIA3395-96
New Product
Rat FGF6 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat FGF6 in serum, plasma or other biological fluids. 1 * 96 Tests
  ANTIA3395-96