ContactUsLinkComponent
Mouse LGI3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f mouse LGI3 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA5103-48
New Product
Mouse LGI3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f mouse LGI3 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA5103-48