ContactUsLinkComponent
Human Collagen X ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human Collagen X in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA4810-96
Supplier:  ANTIBODIES.COM
New Product
Human Collagen X ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human Collagen X in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA4810-96
Supplier:  ANTIBODIES.COM