ContactUsLinkComponent
Rat MIP-1 alpha/CCL3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of rat MIP-1 alpha/CCL3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other 1 * 48 Tests
Catalog # ANTIA3463-48
Supplier:  ANTIBODIES.COM
New Product
Rat MIP-1 alpha/CCL3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of rat MIP-1 alpha/CCL3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other 1 * 48 Tests
Catalog # ANTIA3463-48
Supplier:  ANTIBODIES.COM