ContactUsLinkComponent
Rat Adiponectin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of rat Adiponectin in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3360-96
Supplier:  ANTIBODIES.COM
New Product
Rat Adiponectin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of rat Adiponectin in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3360-96
Supplier:  ANTIBODIES.COM