ContactUsLinkComponent
Human Serotonin Transporter ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitativ e determination of human Serotonin Transporter in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5055-96
Supplier:  ANTIBODIES.COM
New Product
Human Serotonin Transporter ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitativ e determination of human Serotonin Transporter in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5055-96
Supplier:  ANTIBODIES.COM