ContactUsLinkComponent
Mouse Glucagon Like Peptide 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of mouse Glucagon Like Peptide 1 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3615-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Glucagon Like Peptide 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of mouse Glucagon Like Peptide 1 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3615-96
Supplier:  ANTIBODIES.COM