ContactUsLinkComponent
Erythromycin + Lincomycin + Tylosin + Gentamycin Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitati ve determination of Erythromycin + Lincomycin + Tylosin + Gentamycin in raw commingled cow&#0146 s milk. 1 * 96 Tests
Catalog # ANTIA327204-96
Supplier:  ANTIBODIES.COM
New Product
Erythromycin + Lincomycin + Tylosin + Gentamycin Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitati ve determination of Erythromycin + Lincomycin + Tylosin + Gentamycin in raw commingled cow&#0146 s milk. 1 * 96 Tests
Catalog # ANTIA327204-96
Supplier:  ANTIBODIES.COM