ContactUsLinkComponent
Rat Neurotensin ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determi nation of rat Neurotensin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biolog 1 * 96 Tests
  ANTIA4985-96
New Product
Rat Neurotensin ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determi nation of rat Neurotensin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biolog 1 * 96 Tests
  ANTIA4985-96