ContactUsLinkComponent
Rat Osteoprotegerin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of rat Osteoprotegerin in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3447-96
Supplier:  ANTIBODIES.COM
New Product
Rat Osteoprotegerin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of rat Osteoprotegerin in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3447-96
Supplier:  ANTIBODIES.COM