ContactUsLinkComponent
Human Apolipoprotein B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of human Apolipoprotein B in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or o 1 * 96 Tests
Catalog # ANTIA4595-96
Supplier:  ANTIBODIES.COM
New Product
Human Apolipoprotein B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of human Apolipoprotein B in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or o 1 * 96 Tests
Catalog # ANTIA4595-96
Supplier:  ANTIBODIES.COM