ContactUsLinkComponent
Human GRB10 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human GRB10 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA5847-48
New Product
Human GRB10 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human GRB10 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA5847-48