ContactUsLinkComponent
Mouse Peptide YY/PYY ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of mouse Peptide YY/PYY in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other 1 * 48 Tests
Catalog # ANTIA5344-48
Supplier:  Antibodies.com
New Product
Mouse Peptide YY/PYY ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of mouse Peptide YY/PYY in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other 1 * 48 Tests
Catalog # ANTIA5344-48
Supplier:  Antibodies.com