ContactUsLinkComponent
Bovine IL-15 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of bovine IL-15 in serum, plasma or other biological fluids. 1 * 96 Tests
  ANTIA4169-96
New Product
Bovine IL-15 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of bovine IL-15 in serum, plasma or other biological fluids. 1 * 96 Tests
  ANTIA4169-96