ContactUsLinkComponent
Rat Fetuin B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Fetuin B in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3789-96
Supplier:  ANTIBODIES.COM
New Product
Rat Fetuin B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Fetuin B in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3789-96
Supplier:  ANTIBODIES.COM