ContactUsLinkComponent
Rabbit Insulin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of rab bit Insulin in serum, plasma, and other biological fluids. 1 * 96 Tests
Catalog # ANTIA327124-96
Supplier:  ANTIBODIES.COM
New Product
Rabbit Insulin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of rab bit Insulin in serum, plasma, and other biological fluids. 1 * 96 Tests
Catalog # ANTIA327124-96
Supplier:  ANTIBODIES.COM