Mouse Calcium Pump PMCA2 ATPase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of mouse Calcium Pump PMCA2 ATPase in tissue homogenates, cell lysates or other biological fluids 1 * 48 Tests
Catalog # ANTIA4938-48
Supplier: Antibodies.com