ContactUsLinkComponent
Porcine Hepcidin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of porcine Hepcidin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologi 1 * 48 Tests
  ANTIA4842-48
New Product
Porcine Hepcidin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of porcine Hepcidin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologi 1 * 48 Tests
  ANTIA4842-48