ContactUsLinkComponent
Mouse BRCA2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse BRCA2 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5180-48
Supplier:  Antibodies.com
New Product
Mouse BRCA2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse BRCA2 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5180-48
Supplier:  Antibodies.com