ContactUsLinkComponent
Rat MAG/GMA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat MAG/GMA in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA5188-48
New Product
Rat MAG/GMA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat MAG/GMA in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA5188-48