ContactUsLinkComponent
Rat Cytokeratin 18 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of rat Cytokeratin 18 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA4022-96
Supplier:  Antibodies.com
New Product
Rat Cytokeratin 18 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of rat Cytokeratin 18 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA4022-96
Supplier:  Antibodies.com