ContactUsLinkComponent
Mouse Urocortin 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of mouse Urocortin 3 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4947-48
Supplier:  Antibodies.com
New Product
Mouse Urocortin 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of mouse Urocortin 3 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4947-48
Supplier:  Antibodies.com