Canine NT-ProBNP ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determ ination of canine NT-ProBNP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biol 1 * 48 Tests
Catalog # ANTIA4612-48
Supplier: ANTIBODIES.COM