ContactUsLinkComponent
Mouse Vasopressin ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative deter mination of mouse Vasopressin in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5483-48
Supplier:  Antibodies.com
New Product
Mouse Vasopressin ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative deter mination of mouse Vasopressin in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5483-48
Supplier:  Antibodies.com