ContactUsLinkComponent
Human Syndecan 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human Syndecan 3 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5948-48
Supplier:  ANTIBODIES.COM
New Product
Human Syndecan 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human Syndecan 3 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5948-48
Supplier:  ANTIBODIES.COM