ContactUsLinkComponent
Human Sortilin/NT3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of human Sortilin/NT3 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5206-96
Supplier:  ANTIBODIES.COM
New Product
Human Sortilin/NT3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of human Sortilin/NT3 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5206-96
Supplier:  ANTIBODIES.COM