ContactUsLinkComponent
Mouse Cholecystokinin 8 ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of mouse Cholecystokinin 8 in serum, plasma, tissue homogenates, cell culture supernates or other biol 1 * 96 Tests
Catalog # ANTIA3808-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Cholecystokinin 8 ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of mouse Cholecystokinin 8 in serum, plasma, tissue homogenates, cell culture supernates or other biol 1 * 96 Tests
Catalog # ANTIA3808-96
Supplier:  ANTIBODIES.COM