ContactUsLinkComponent
Human HHIP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human HHIP in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5518-48
Supplier:  ANTIBODIES.COM
New Product
Human HHIP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human HHIP in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5518-48
Supplier:  ANTIBODIES.COM