ContactUsLinkComponent
Human Fibrinogen gamma Chain ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitati ve determination of human Fibrinogen gamma Chain in plasma. 1 * 48 Tests
Catalog # ANTIA5417-48
Supplier:  ANTIBODIES.COM
New Product
Human Fibrinogen gamma Chain ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitati ve determination of human Fibrinogen gamma Chain in plasma. 1 * 48 Tests
Catalog # ANTIA5417-48
Supplier:  ANTIBODIES.COM