ContactUsLinkComponent
Rat Parvalbumin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of rat Parvalbumin in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5671-48
Supplier:  Antibodies.com
New Product
Rat Parvalbumin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of rat Parvalbumin in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5671-48
Supplier:  Antibodies.com