ContactUsLinkComponent
Human HLA C ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human HLA C in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4582-48
Supplier:  Antibodies.com
New Product
Human HLA C ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human HLA C in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4582-48
Supplier:  Antibodies.com