ContactUsLinkComponent
Mouse TREM1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse TREM1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA4059-96
Supplier:  ANTIBODIES.COM
New Product
Mouse TREM1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse TREM1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA4059-96
Supplier:  ANTIBODIES.COM