ContactUsLinkComponent
Human ATG16L1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human ATG16L1 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5906-48
Supplier:  Antibodies.com
New Product
Human ATG16L1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human ATG16L1 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5906-48
Supplier:  Antibodies.com