ContactUsLinkComponent
Mouse Ryanodine Receptor ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of mouse Ryanodine Receptor in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5270-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Ryanodine Receptor ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of mouse Ryanodine Receptor in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5270-96
Supplier:  ANTIBODIES.COM