ContactUsLinkComponent
Mouse SFRP5 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse SFRP5 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4987-48
Supplier:  Antibodies.com
New Product
Mouse SFRP5 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse SFRP5 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4987-48
Supplier:  Antibodies.com