ContactUsLinkComponent
Mouse Progesterone ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of mouse Progesterone in serum and plasma. 1 * 96 Tests
Catalog # ANTIA327143-96
Supplier:  Antibodies.com
New Product
Mouse Progesterone ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of mouse Progesterone in serum and plasma. 1 * 96 Tests
Catalog # ANTIA327143-96
Supplier:  Antibodies.com