ContactUsLinkComponent
Human Desmocollin 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of human Desmocollin 3 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5467-96
Supplier:  Antibodies.com
New Product
Human Desmocollin 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of human Desmocollin 3 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5467-96
Supplier:  Antibodies.com