ContactUsLinkComponent
Chicken CD30-L ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of chicken CD30-L in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4007-48
Supplier:  Antibodies.com
New Product
Chicken CD30-L ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of chicken CD30-L in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4007-48
Supplier:  Antibodies.com