ContactUsLinkComponent
Human BTN1A1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human BTN1A1 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5758-48
Supplier:  Antibodies.com
New Product
Human BTN1A1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human BTN1A1 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5758-48
Supplier:  Antibodies.com