ContactUsLinkComponent
Mouse SerpinB4/SCCA-2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of mouse SerpinB4/SCCA-2 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4196-48
Supplier:  Antibodies.com
New Product
Mouse SerpinB4/SCCA-2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of mouse SerpinB4/SCCA-2 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4196-48
Supplier:  Antibodies.com