ContactUsLinkComponent
Human KIF1A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human KIF1A in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5088-96
Supplier:  ANTIBODIES.COM
New Product
Human KIF1A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human KIF1A in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5088-96
Supplier:  ANTIBODIES.COM