ContactUsLinkComponent
Mouse ICAM2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse ICAM2 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3527-48
Supplier:  Antibodies.com
New Product
Mouse ICAM2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse ICAM2 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3527-48
Supplier:  Antibodies.com