ContactUsLinkComponent
Human PC1/3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human PC1/3 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5086-96
Supplier:  Antibodies.com
New Product
Human PC1/3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human PC1/3 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5086-96
Supplier:  Antibodies.com