ContactUsLinkComponent
Human SLC1A5/ASCT2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of human SLC1A5/ASCT2 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA5609-48
New Product
Human SLC1A5/ASCT2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of human SLC1A5/ASCT2 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA5609-48