ContactUsLinkComponent
Mouse Complement Factor B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Complement Factor B in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3999-48
Supplier:  ANTIBODIES.COM
New Product
Mouse Complement Factor B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Complement Factor B in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3999-48
Supplier:  ANTIBODIES.COM