ContactUsLinkComponent
Human RIOK1/RIO1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human RIOK1/RIO1 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5459-96
Supplier:  Antibodies.com
New Product
Human RIOK1/RIO1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human RIOK1/RIO1 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5459-96
Supplier:  Antibodies.com