ContactUsLinkComponent
Mouse Cyclophilin B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of mouse Cyclophilin B in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4289-48
Supplier:  Antibodies.com
New Product
Mouse Cyclophilin B ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of mouse Cyclophilin B in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4289-48
Supplier:  Antibodies.com