ContactUsLinkComponent
Human Dhh ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Dhh in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5400-96
Supplier:  Antibodies.com
New Product
Human Dhh ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Dhh in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5400-96
Supplier:  Antibodies.com