ContactUsLinkComponent
Human PI 3 Kinase p110 beta ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitativ e determination of human PI 3 Kinase p110 beta in tissue homogenates or other biological fluids. 1 * 96 Tests
  ANTIA5918-96
New Product
Human PI 3 Kinase p110 beta ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitativ e determination of human PI 3 Kinase p110 beta in tissue homogenates or other biological fluids. 1 * 96 Tests
  ANTIA5918-96