ContactUsLinkComponent
Mouse PDGFC ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse PDGFC in plasma. 1 * 96 Tests
Catalog # ANTIA5479-96
Supplier:  Antibodies.com
New Product
Mouse PDGFC ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse PDGFC in plasma. 1 * 96 Tests
Catalog # ANTIA5479-96
Supplier:  Antibodies.com